Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008895.2 | NP_001008895.1 | 16 | 1784 | ||
NM_001278513.1 | NP_001265442.1 | 16 | 1928 | |||
NM_001278514.1 | NP_001265443.1 | 16 | 1596 | |||
NM_003589.2 | NP_003580.1 | 16 | 1610 | |||
XM_011537523.2 | XP_011535825.1 | 15 | 2144 | |||
GenBank mRNA | AB012193.1 | - | 10 | 958 | ||
AB178950.1 | - | 16 | 1661 | |||
AF077188.1 | - | 16 | 1521 | |||
AK296700.1 | - | 8 | 905 | |||
AK314835.1 | - | 16 | 1883 | |||
AL833355.1 | - | 15 | 1548 | |||
AY365124.1 | - | 16 | 1661 | |||
BC008308.1 | - | 16 | 1672 | 3537176 | ||
U58090.1 | - | 6 | 648 |