Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sord | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017052.2 | NP_058748.2 | 7 | 727 | ||
GenBank mRNA | BC088398.1 | - | 7 | 727 | 7112546 | |
BC098919.1 | - | 7 | 674 | 7373210 | ||
BC128707.1 | - | 7 | 703 | 8373758 | ||
FQ209461.1 | - | 7 | 722 | |||
FQ210296.1 | - | 6 | 1887 | |||
FQ211554.1 | - | 5 | 1764 | |||
FQ234001.1 | - | 7 | 616 | |||
X59037.1 | - | 7 | 667 |
Gene Symbol: Usp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY387057.1 | - | 2 | 590 |