Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017069.3 | NP_058765.3 | 7, 8 | 1023 | ||
XM_006229511.2 | XP_006229573.1 | 7, 8 | 1135 | |||
GenBank mRNA | CB725341.1 | - | 1, 2 | 59 | ||
X51991.1 | - | 7, 8 | 1030 |