Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256658.1 | NP_001243587.1 | 12 | 2203 | ||
NM_001256659.1 | NP_001243588.1 | 12 | 2159 | |||
NM_001256660.1 | NP_001243589.1 | 13 | 2202 | |||
NM_001256661.1 | NP_001243590.1 | 13 | 2177 | |||
NM_001256662.1 | NP_001243591.1 | 12 | 1933 | |||
NM_003598.1 | NP_003589.1 | 12 | 2150 | |||
XM_005259334.3 | XP_005259391.1 | 10 | 2314 | |||
XM_006723424.1 | XP_006723487.1 | 12 | 2434 | |||
XM_006723428.2 | XP_006723491.1 | 11 | 2383 | |||
XM_006723429.2 | XP_006723492.1 | 7 | 1635 | |||
XM_011527398.1 | XP_011525700.1 | 12 | 2443 | |||
XM_011527399.2 | XP_011525701.1 | 13 | 2166 | |||
XM_011527400.2 | XP_011525702.1 | 13 | 2136 | |||
XM_011527401.1 | XP_011525703.1 | 13 | 2109 | |||
XM_011527402.2 | XP_011525704.1 | 13 | 2111 | |||
XM_011527403.1 | XP_011525705.1 | 12 | 2135 | |||
XM_011527404.1 | XP_011525706.1 | 12 | 2404 | |||
GenBank mRNA | AL832240.1 | - | 13 | 2126 | ||
BC007556.1 | - | 12 | 2150 | 2967735 | ||
BC018803.1 | - | 13 | 2176 | |||
BC051301.1 | - | 12 | 2154 | 4843913 |