Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017101.1 | NP_058797.1 | 3 | 197 | ||
GenBank mRNA | BC059141.1 | - | 3 | 202 | 6919594 | |
BC091153.1 | - | 3 | 173 | 7308693 | ||
FQ210268.1 | - | 3 | 198 | |||
FQ210843.1 | - | 3 | 200 | |||
FQ222826.1 | - | 3 | 198 | |||
FQ223366.1 | - | 3 | 198 | |||
FQ223500.1 | - | 3 | 198 | |||
FQ227889.1 | - | 3 | 198 | |||
FQ228411.1 | - | 3 | 198 | |||
FQ228450.1 | - | 3 | 198 | |||
FQ231674.1 | - | 3 | 198 | |||
FQ233894.1 | - | 3 | 197 | |||
FQ234872.1 | - | 3 | 198 | |||
M19533.1 | - | 3 | 197 | |||
M25637.1 | - | 3 | 190 |
Gene Symbol: LOC100360977 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250801.2 | XP_006250863.1 | 1 | 194 |
Gene Symbol: RGD1559682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_341363.4 | XP_341364.2 | 1 | 155 |