Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017109.1 | NP_058805.1 | 1 | 400 | ||
GenBank mRNA | AF056704.1 | - | 1 | 400 |
Gene Symbol: LOC100910134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765288.1 | XP_008763510.1 | 2 | 695 | ||
XM_008765289.1 | XP_008763511.1 | 3 | 798 | |||
XM_008765290.1 | XP_008763512.1 | 2 | 671 |