Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017122.1 | NP_058818.1 | 1, 2 | 560 | ||
XM_006238920.1 | XP_006238982.1 | 2, 3 | 505 | |||
XM_006238921.2 | XP_006238983.1 | 2, 3 | 462 | |||
GenBank mRNA | AY442172.1 | - | 1, 2 | 387 | ||
BC087632.1 | - | 1, 2 | 415 | 7319524 | ||
D12573.1 | - | 1, 2 | 560 |