Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017125.3 | NP_058821.1 | 5, 6 | 491 | ||
XM_008765024.1 | XP_008763246.1 | 5, 6 | 553 | |||
XM_008765025.1 | XP_008763247.1 | 5, 6 | 756 | |||
GenBank mRNA | AF159104.1 | - | 1, 2 | 115 | ||
BC063173.1 | - | 5, 6 | 494 | 6921594 | ||
FQ214468.1 | - | 5, 6 | 558 | |||
FQ221333.1 | - | 5, 6 | 495 | |||
FQ222038.1 | - | 5, 6 | 474 | |||
FQ230116.1 | - | 5, 6 | 462 | |||
X61654.1 | - | 5, 6 | 491 |