Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inhba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017128.2 | NP_058824.1 | 2 | 346 | ||
XM_006254001.2 | XP_006254063.1 | 3 | 4005 | |||
XM_008771712.1 | XP_008769934.1 | 2 | 3923 | |||
XM_008771713.1 | XP_008769935.1 | 4 | 5713 | |||
XM_008771714.1 | XP_008769936.1 | 4 | 2493 | |||
XM_008771715.1 | XP_008769937.1 | 4 | 2395 | |||
XM_008771716.1 | XP_008769938.1 | 4 | 773 | |||
GenBank mRNA | M37482.1 | - | 2 | 338 |