Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nefm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017029.2 | NP_058725.1 | 3 | 2221 | ||
XM_008770736.1 | XP_008768958.1 | 4 | 2558 | |||
GenBank mRNA | M18628.1 | - | 3 | 2196 | ||
Z12152.1 | - | 3 | 2224 |