Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017039.2 | NP_058735.1 | 2 | 473 | ||
GenBank mRNA | BC070914.1 | - | 2 | 482 | 7104697 | |
BC072531.1 | - | 2 | 473 | 7096821 | ||
FQ228785.1 | - | 1 | 37 | |||
M33114.1 | - | 2 | 389 | |||
X14159.1 | - | 2 | 389 | |||
X16043.1 | - | 2 | 448 |
Gene Symbol: LOC103694903 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773894.1 | XP_008772116.1 | 968 |