Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BARX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003658.4 | NP_003649.2 | 4 | 938 | ||
XM_011543043.1 | XP_011541345.1 | 4 | 765 | |||
XM_011543044.2 | XP_011541346.1 | 4 | 783 | |||
GenBank mRNA | AF031924.1 | - | 4 | 982 | ||
AJ243512.1 | - | 4 | 904 |