Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017127.1 | NP_058823.1 | 4 | 932 | ||
XM_006230707.2 | XP_006230769.1 | 6 | 1955 | |||
XM_006230708.2 | XP_006230770.1 | 5 | 1924 | |||
XM_008760090.1 | XP_008758312.1 | 4 | 3247 | |||
XM_008760091.1 | XP_008758313.1 | 5 | 3407 | |||
GenBank mRNA | BC081821.1 | - | 4 | 908 | 7109261 | |
D10262.1 | - | 4 | 932 | |||
D37885.1 | - | 5 | 688 |