Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017160.1 | NP_058856.1 | 161 | |||
GenBank mRNA | BC058149.1 | - | 191 | 6918741 | ||
M29358.1 | - | 161 |
Gene Symbol: LOC100911372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749971.3 | XP_003750019.1 | 2, 3 | 246 |