Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035233.1 | NP_001030310.1 | 379 | |||
NM_017215.2 | NP_058911.2 | 379 | ||||
GenBank mRNA | AF297648.1 | - | 291 | |||
AF465909.1 | - | 272 | ||||
AY044832.1 | - | 268 | ||||
CB760876.1 | - | 380 | ||||
JN132400.1 | - | 673 | ||||
U15098.1 | - | 328 | ||||
X67857.1 | - | 366 |