Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017224.2 | NP_058920.1 | 3 | 542 | ||
XM_006230978.2 | XP_006231040.1 | 3 | 797 | |||
GenBank mRNA | AB004559.1 | - | 3 | 772 | ||
AF008221.1 | - | 3 | 750 | |||
BC104692.1 | - | 3 | 542 | 7109188 |