Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPSA SIRNA Assays ›
Gene Symbol:
Rpsa
Gene Name:
ribosomal protein SA
Gene Aliases:
Lamr1
Chromosome Location:
Chr.8: 128806056 - 128809987 on Build Rnor_6.0
UniGene ID:
Rn.121972
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
49914
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rpsa
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017138.2
NP_058834.1
3
230
XM_008766672.1
XP_008764894.1
3
303
GenBank mRNA
BC060578.1
-
3
248
6919235
D25224.1
-
3
231
FJ386454.1
-
2
211
FQ209463.1
-
3
244
FQ209469.1
-
3
244
FQ209490.1
-
3
245
FQ209667.1
-
3
244
FQ209729.1
-
3
244
FQ211189.1
-
3
244
FQ212660.1
-
3
244
FQ212893.1
-
3
244
FQ213275.1
-
3
244
FQ213317.1
-
3
244
FQ213334.1
-
3
244
FQ213623.1
-
3
247
FQ213835.1
-
3
245
FQ215242.1
-
3
244
FQ215292.1
-
3
245
FQ216063.1
-
3
244
FQ216085.1
-
3
244
FQ216130.1
-
3
244
FQ216245.1
-
3
244
FQ216788.1
-
3
246
FQ216997.1
-
3
244
FQ217664.1
-
3
244
FQ219586.1
-
3
244
FQ219785.1
-
3
247
FQ220012.1
-
3
244
FQ220017.1
-
3
242
FQ220075.1
-
3
245
FQ220084.1
-
3
247
FQ220126.1
-
3
244
FQ220134.1
-
3
244
FQ220171.1
-
3
243
FQ220184.1
-
3
244
FQ220347.1
-
3
246
FQ220461.1
-
3
225
FQ220468.1
-
3
244
FQ220661.1
-
3
247
FQ220822.1
-
3
244
FQ220904.1
-
3
245
FQ220964.1
-
3
244
FQ221103.1
-
3
244
FQ221334.1
-
3
264
FQ221443.1
-
3
245
FQ221672.1
-
3
244
FQ222093.1
-
3
246
FQ222280.1
-
3
244
FQ222542.1
-
3
246
FQ223223.1
-
3
244
FQ224747.1
-
3
244
FQ225510.1
-
3
245
FQ226054.1
-
3
244
FQ226408.1
-
3
244
FQ227056.1
-
3
244
FQ227293.1
-
3
244
FQ227600.1
-
3
245
FQ228002.1
-
3
244
FQ228212.1
-
3
245
FQ229290.1
-
3
246
FQ229305.1
-
3
246
FQ229367.1
-
3
245
FQ229516.1
-
3
244
FQ229579.1
-
3
244
FQ229598.1
-
3
245
FQ229712.1
-
3
244
FQ229787.1
-
3
244
FQ229850.1
-
3
246
FQ229890.1
-
3
244
FQ229918.1
-
3
264
FQ229970.1
-
3
244
FQ230088.1
-
3
244
FQ230180.1
-
3
245
FQ230258.1
-
3
243
FQ231469.1
-
2
61
FQ231622.1
-
3
242
FQ232305.1
-
3
246
FQ232568.1
-
3
245
FQ233068.1
-
3
244
FQ233112.1
-
3
244
FQ233340.1
-
3
246
FQ233547.1
-
3
245
FQ233919.1
-
3
244
FQ234511.1
-
3
244
FQ234742.1
-
3
244
FQ234765.1
-
3
244
FQ234772.1
-
3
244
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
Gene Ontology Categories:
Function(s)
Process(es)
ribosomal small subunit assembly
endonucleolytic cleavage in ITS1 to separate SSU-rRNA from 5.8S rRNA and LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
endonucleolytic cleavage to generate mature 3'-end of SSU-rRNA from (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
rRNA export from nucleus
translation
single organismal cell-cell adhesion
epithelial cell differentiation
structural constituent of ribosome
laminin receptor activity
structural molecule activity
ribosome binding
laminin binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline