Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNGTT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286426.1 | NP_001273355.1 | 15 | 3631 | ||
NM_001286428.1 | NP_001273357.1 | 14 | 3521 | |||
NM_003800.4 | NP_003791.3 | 16 | 3700 | |||
XM_017011401.1 | XP_016866890.1 | 17 | 3805 | |||
GenBank mRNA | AF025654.1 | - | 16 | 3769 | ||
BC019954.1 | - | 16 | 3667 | 3923156 | ||
BX537450.1 | - | 3 | 4888 |