Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017147.2 | NP_058843.1 | 2 | 248 | ||
GenBank mRNA | BC059143.1 | - | 2 | 248 | 6889053 | |
BC086533.1 | - | 2 | 233 | 7305422 | ||
FQ212136.1 | - | 1 | 121 | |||
FQ212794.1 | - | 2 | 247 | |||
FQ212910.1 | - | 2 | 243 | |||
FQ213701.1 | - | 2 | 243 | |||
FQ213897.1 | - | 2 | 272 | |||
FQ214120.1 | - | 2 | 244 | |||
FQ214438.1 | - | 2 | 243 | |||
FQ220384.1 | - | 2 | 259 | |||
FQ220571.1 | - | 2 | 245 | |||
FQ226405.1 | - | 2 | 243 | |||
FQ227487.1 | - | 2 | 245 | |||
FQ229073.1 | - | 2 | 272 | |||
FQ229766.1 | - | 2 | 247 | |||
FQ229833.1 | - | 2 | 243 | |||
FQ229875.1 | - | 2 | 247 | |||
FQ230001.1 | - | 2 | 247 | |||
FQ230091.1 | - | 2 | 244 | |||
FQ231379.1 | - | 2 | 243 | |||
FQ232577.1 | - | 1 | 123 | |||
FQ233750.1 | - | 2 | 243 | |||
FQ234162.1 | - | 2 | 247 | |||
FQ234303.1 | - | 2 | 243 | |||
X62908.1 | - | 1 | 172 |