Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017170.2 | NP_058866.2 | 2 | 473 | ||
XM_008769786.1 | XP_008768008.1 | 3 | 782 | |||
GenBank mRNA | M83177.1 | - | 2 | 473 | ||
X55761.1 | - | 2 | 555 |