Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083586.2 | NP_001077055.2 | 2, 3 | 489 | ||
NM_017197.3 | NP_058893.2 | 2, 3 | 489 | |||
XM_008771861.1 | XP_008770083.1 | 2, 3 | 368 | |||
XM_008771862.1 | XP_008770084.1 | 1, 2 | 926 | |||
XM_008771863.1 | XP_008770085.1 | 1, 2 | 925 | |||
XM_008771864.1 | XP_008770086.1 | 1, 2 | 925 | |||
GenBank mRNA | AF090695.1 | - | 2, 3 | 323 | ||
AJ010351.1 | - | 2, 3 | 414 | |||
AJ010386.1 | - | 2, 3 | 537 | |||
BC129096.1 | - | 2, 3 | 315 | 8362847 |