Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017204.1 | NP_058900.1 | 2 | 1905 | ||
XM_006229755.2 | XP_006229817.1 | 1 | 2235 | |||
GenBank mRNA | AJ002556.1 | - | 1 | 1924 | ||
BC078848.1 | - | 1 | 1783 | |||
X93495.1 | - | 2 | 1905 |