Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017209.1 | NP_058905.1 | 2, 3 | 301 | ||
XM_006239009.2 | XP_006239071.1 | 3, 4 | 420 | |||
GenBank mRNA | AB015724.1 | - | 2, 3 | 301 | ||
FQ210707.1 | - | 2, 3 | 297 |