Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306144.1 | NP_001293073.1 | 16 | 4307 | ||
NM_003828.3 | NP_003819.1 | 16 | 2509 | |||
XM_005274765.3 | XP_005274822.1 | 18 | 4347 | |||
XM_005274766.3 | XP_005274823.1 | 17 | 4295 | |||
XM_005274767.3 | XP_005274824.1 | 16 | 4314 | |||
XM_006724855.3 | XP_006724918.1 | 15 | 4283 | |||
XM_011531209.2 | XP_011529511.1 | 15 | 4043 | |||
XM_017029921.1 | XP_016885410.1 | 16 | 4078 | |||
XM_017029922.1 | XP_016885411.1 | 16 | 4090 | |||
XM_017029923.1 | XP_016885412.1 | 13 | 3870 | |||
GenBank mRNA | AF057354.1 | - | 17 | 2544 | ||
AJ224979.1 | - | 16 | 2334 | |||
AK001816.1 | - | 13 | 1889 | |||
AK024745.1 | - | 1 | 989 | |||
AK055513.1 | - | 1 | 2051 | |||
AK226109.1 | - | 16 | 4260 | |||
BC011250.1 | - | 1 | 1938 |