Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pctp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017225.2 | NP_058921.1 | 4 | 391 | ||
XM_006247124.2 | XP_006247186.1 | 4 | 377 | |||
XM_006247125.2 | XP_006247187.1 | 4 | 459 | |||
XM_008767982.1 | XP_008766204.1 | 6 | 598 | |||
XM_008767983.1 | XP_008766205.1 | 6 | 552 | |||
GenBank mRNA | AF040261.1 | - | 4 | 335 | ||
BC078854.1 | - | 4 | 396 | 7113616 | ||
Z50025.1 | - | 2 | 94 |