Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017229.1 | NP_058925.1 | 3 | 1198 | ||
GenBank mRNA | Z22867.1 | - | 3 | 1198 |