Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitpna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017231.1 | NP_058927.1 | 3 | 366 | ||
GenBank mRNA | BC070945.1 | - | 3 | 394 | 7103390 | |
M25758.1 | - | 3 | 366 |