Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm14295 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205057.2 | NP_001191986.1 | 1263 | |||
GenBank mRNA | BC083167.1 | - | 1216 |
Gene Symbol: 0610010B08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002444.1 | - | 4 | 1249 | ||
BC099528.1 | - | 4 | 1223 |
Gene Symbol: Gm14325 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC092391.1 | - | 3 | 1188 | 6853907 |
Gene Symbol: Gm14399 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK166773.1 | - | 1162 |
Gene Symbol: Gm14419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC025129.1 | - | 3 | 1138 | ||
BC085231.1 | - | 4 | 1238 |
Gene Symbol: LOC628147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005481.1 | - | 5 | 676 |