Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm14295 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205057.2 | NP_001191986.1 | 1072 | |||
GenBank mRNA | BC083167.1 | - | 1025 | |||
BC106197.1 | - | 1056 |
Gene Symbol: 0610010B08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002444.1 | - | 4 | 1056 | ||
BC099528.1 | - | 4 | 1031 |
Gene Symbol: Gm14305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC081427.1 | - | 4 | 1023 | ||
BC100574.1 | - | 4 | 1045 |
Gene Symbol: Gm14326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC042711.1 | - | 4 | 988 |
Gene Symbol: Gm14399 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK166773.1 | - | 969 |
Gene Symbol: Gm14419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC025129.1 | - | 3 | 946 | ||
BC085231.1 | - | 4 | 1046 |
Gene Symbol: LOC628147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005481.1 | - | 5 | 484 |