Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPSA SIRNA Assays ›
Gene Symbol:
Rpsa
Gene Name:
ribosomal protein SA
Gene Aliases:
Lamr1
Chromosome Location:
Chr.8: 128806056 - 128809987 on Build Rnor_6.0
UniGene ID:
Rn.121972
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
50006
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rpsa
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017138.2
NP_058834.1
5
660
XM_008766672.1
XP_008764894.1
5
733
GenBank mRNA
BC060578.1
-
5
678
6919235
D25224.1
-
5
661
FJ386454.1
-
4
641
FQ209469.1
-
5
674
FQ209490.1
-
5
675
FQ209667.1
-
5
674
FQ209729.1
-
5
675
FQ212660.1
-
5
674
FQ212893.1
-
5
674
FQ213275.1
-
5
674
FQ213317.1
-
5
673
FQ213334.1
-
5
674
FQ213623.1
-
5
677
FQ213835.1
-
5
675
FQ213894.1
-
5
674
FQ215242.1
-
5
674
FQ215292.1
-
5
675
FQ216063.1
-
5
674
FQ216085.1
-
5
674
FQ216130.1
-
5
674
FQ216245.1
-
5
674
FQ216788.1
-
5
676
FQ216997.1
-
5
674
FQ217664.1
-
5
674
FQ219586.1
-
5
674
FQ219785.1
-
5
677
FQ220012.1
-
5
674
FQ220017.1
-
5
672
FQ220075.1
-
5
675
FQ220084.1
-
5
677
FQ220126.1
-
5
674
FQ220134.1
-
5
674
FQ220171.1
-
5
673
FQ220184.1
-
5
674
FQ220347.1
-
5
676
FQ220461.1
-
5
655
FQ220468.1
-
5
674
FQ220661.1
-
5
677
FQ220822.1
-
5
674
FQ220904.1
-
5
675
FQ220964.1
-
5
674
FQ221103.1
-
5
674
FQ221334.1
-
5
694
FQ221443.1
-
5
675
FQ221672.1
-
5
674
FQ222093.1
-
5
676
FQ222280.1
-
5
674
FQ222542.1
-
5
676
FQ222757.1
-
5
672
FQ223223.1
-
5
674
FQ224747.1
-
5
674
FQ225510.1
-
5
675
FQ226054.1
-
5
674
FQ226408.1
-
5
674
FQ227056.1
-
5
674
FQ227293.1
-
5
674
FQ227600.1
-
5
676
FQ228002.1
-
5
674
FQ228212.1
-
5
675
FQ229290.1
-
5
676
FQ229305.1
-
5
676
FQ229367.1
-
5
675
FQ229516.1
-
5
674
FQ229579.1
-
5
674
FQ229598.1
-
5
675
FQ229712.1
-
5
674
FQ229787.1
-
5
674
FQ229850.1
-
5
676
FQ229859.1
-
5
672
FQ229890.1
-
5
674
FQ229918.1
-
5
694
FQ229945.1
-
5
674
FQ229970.1
-
5
674
FQ230088.1
-
5
674
FQ230106.1
-
5
674
FQ230180.1
-
5
675
FQ230258.1
-
5
673
FQ231469.1
-
4
491
FQ231622.1
-
5
672
FQ232305.1
-
5
676
FQ232568.1
-
5
675
FQ233068.1
-
5
673
FQ233112.1
-
5
674
FQ233340.1
-
5
676
FQ233547.1
-
5
675
FQ233919.1
-
5
675
FQ234511.1
-
5
674
FQ234742.1
-
5
674
FQ234765.1
-
5
674
FQ234772.1
-
5
674
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
Gene Ontology Categories:
Function(s)
Process(es)
ribosomal small subunit assembly
endonucleolytic cleavage in ITS1 to separate SSU-rRNA from 5.8S rRNA and LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
endonucleolytic cleavage to generate mature 3'-end of SSU-rRNA from (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
rRNA export from nucleus
translation
single organismal cell-cell adhesion
epithelial cell differentiation
structural constituent of ribosome
laminin receptor activity
structural molecule activity
ribosome binding
laminin binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline