Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm6182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK163771.1 | - | 1 | 2143 | ||
AK166086.1 | - | 5 | 3263 | |||
BC048160.1 | - | 5 | 3227 | |||
BC051922.1 | - | 5 | 3227 | |||
BC059210.1 | - | 4 | 4361 |