Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ildr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164528.1 | NP_001158000.1 | 10 | 6570 | ||
GenBank mRNA | AK013417.1 | - | 1 | 41 |