Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017150.1 | NP_058846.1 | 4 | 426 | ||
GenBank mRNA | BC086404.1 | - | 4 | 420 | 7304319 | |
FQ221016.1 | - | 4 | 427 | |||
FQ221647.1 | - | 4 | 433 | |||
FQ222035.1 | - | 4 | 448 | |||
FQ229056.1 | - | 4 | 431 | |||
FQ229057.1 | - | 4 | 431 | |||
X60744.1 | - | 4 | 455 | |||
X68283.1 | - | 4 | 426 |
Gene Symbol: RGD1563300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253902.2 | XP_006253964.2 | 2 | 429 |