Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017152.3 | NP_058848.2 | 3 | 273 | ||
GenBank mRNA | BC058484.1 | - | 3 | 277 | 6921184 | |
FQ209589.1 | - | 3 | 270 | |||
FQ210096.1 | - | 3 | 263 | |||
FQ221123.1 | - | 3 | 268 | |||
FQ221680.1 | - | 3 | 263 | |||
FQ222308.1 | - | 3 | 263 | |||
FQ222382.1 | - | 3 | 275 | |||
FQ222858.1 | - | 3 | 268 | |||
FQ223398.1 | - | 3 | 270 | |||
FQ224175.1 | - | 3 | 263 | |||
FQ228331.1 | - | 3 | 268 | |||
FQ228517.1 | - | 3 | 270 | |||
FQ229910.1 | - | 3 | 270 | |||
K02933.1 | - | 3 | 271 |