Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017158.2 | NP_058854.1 | 7, 8 | 1178 | ||
GenBank mRNA | BC097939.1 | - | 7, 8 | 1161 | 7368637 | |
BC127503.1 | - | 7, 8 | 1217 | 7371672 | ||
M18335.1 | - | 7, 8 | 1179 | |||
M31031.1 | - | 7, 8 | 1136 |