Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100606.1 | NP_001094076.1 | 2 | 1098 | ||
GenBank mRNA | AK020747.1 | - | 1 | 755 |