Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017175.2 | NP_058871.2 | 20 | 2657 | ||
XM_006255245.2 | XP_006255307.1 | 20 | 3697 | |||
GenBank mRNA | BC061836.1 | - | 20 | 2558 | 5599989 | |
D26180.1 | - | 20 | 2608 | |||
EV774518.1 | - | 3 | 277 | |||
FQ222217.1 | - | 20 | 2657 | |||
L35634.1 | - | 20 | 2500 |