Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017176.2 | NP_058872.1 | 4 | 489 | ||
GenBank mRNA | AF166100.1 | - | 4 | 491 |