Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195265.1 | NP_001182194.1 | 16 | 3590 | ||
GenBank mRNA | AK033653.1 | - | 6 | 1485 | ||
BC096488.1 | - | 6 | 1706 |