Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc5os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195579.1 | NP_001182508.1 | 4 | 2898 | ||
GenBank mRNA | AK147193.1 | - | 4 | 2899 |