Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK146285.1 | - | 6 | 737 | ||
AK153881.1 | - | 682 | ||||
BC083085.1 | - | 670 | 6516041 | |||
BC084680.1 | - | 6 | 619 | 6477991 | ||
BC092060.1 | - | 6 | 660 | 4020647 | ||
BC103792.1 | - | 666 | 6516212 | |||
X12944.1 | - | 623 |