Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pet100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195244.1 | NP_001182173.1 | 4 | 1122 | ||
GenBank mRNA | AK011549.1 | - | 4 | 1100 |