Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195537.1 | NP_001182466.1 | 3 | 944 | ||
GenBank mRNA | AK005822.1 | - | 2 | 226 | ||
BY705974.1 | - | 2 | 223 | |||
CF197814.1 | - | 1 | 20 | |||
GU144514.1 | - | 3 | 944 |