Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp518b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081144.2 | NP_001074613.2 | 4 | 6237 | ||
NM_001177902.1 | NP_001171373.1 | 3 | 6171 | |||
GenBank mRNA | AK078507.1 | - | 1 | 3485 | ||
AK173245.1 | - | 1 | 5215 |