Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm21950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270667.1 | NP_001257596.1 | 1 | 1246 | ||
GenBank mRNA | AB055855.1 | - | 1 | 1246 |