Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm21951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270669.1 | NP_001257598.1 | 1 | 859 | ||
GenBank mRNA | AB055854.1 | - | 1 | 859 |