Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpusd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033204.1 | NP_001028376.1 | 5, 6 | 518 | ||
GenBank mRNA | AK076749.1 | - | 5, 6 | 519 | ||
AK133871.1 | - | 5, 6 | 498 | |||
BC038081.1 | - | 5, 6 | 459 | |||
BC116825.1 | - | 5, 6 | 518 | 40126144 | ||
BC116827.1 | - | 5, 6 | 518 | 40126146 | ||
BC144733.1 | - | 5, 6 | 518 | 9053281 |