Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081118.1 | NP_001074587.1 | 18 | 5132 | ||
GenBank mRNA | AK042050.1 | - | 5 | 866 | ||
AK129387.1 | - | 3 | 3908 | |||
AK134140.1 | - | 5 | 2701 | |||
AK137120.1 | - | 5 | 798 | |||
AK161749.1 | - | 5 | 2277 | |||
AK164283.1 | - | 5 | 1576 | |||
AK164438.1 | - | 5 | 2050 | |||
BC039628.1 | - | 5 | 890 | |||
BC055395.1 | - | 5 | 951 | |||
BC094566.1 | - | 7 | 3607 | |||
BC138446.1 | - | 13 | 4778 | 8861466 | ||
BC145719.1 | - | 13 | 4778 | 40131022 |