Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pwwp2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098636.1 | NP_001092106.1 | 2 | 1830 | ||
GenBank mRNA | AK213732.1 | - | 1 | 299 |