Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024504.2 | NP_001019675.2 | 5 | 894 | ||
NM_001042651.1 | NP_001036116.1 | 5 | 894 | |||
GenBank mRNA | AK034394.1 | - | 5 | 602 | ||
AK155726.1 | - | 5 | 610 | |||
AK161694.1 | - | 5 | 599 | |||
BC030839.1 | - | 5 | 602 |